Merck
  • Creation of the final abutment before surgery in immediate tooth replacement: two different approaches to obtain a customized zirconia abutment.
Creation of the final abutment before surgery in immediate tooth replacement: two different approaches to obtain a customized zirconia abutment.
The European journal of esthetic dentistry : official journal of the European Academy of Esthetic Dentistry (2013-08-21)
Francesco Mintrone, Ivo Sighinolfi
ABSTRACT
Specialist literature shows that the repeated disconnection of the transmucosal components can generate tissue recession due to the alteration of the delicate environment created between the abutment and surrounding soft tissue. This article will analyze two different techniques of creating the final abutment in the pre-surgery phase, evaluating 2 failing central incisors considered suitable for the immediate tooth replacement technique.
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O