Merck
Modular enantioselective synthesis of 8-aza-prostaglandin E1.
The Journal of organic chemistry (2013-08-21)
Xiao-Gang Wang, Ai-E Wang, Yi Hao, Yuan-Ping Ruan, Pei-Qiang Huang
ABSTRACT
We report herein for the first time the enantioselective synthesis of 8-aza-PGE1. The synthesis used the cross olefin metathesis reaction to connect the 5-vinyl-γ-lactam subunit, prepared from (R)-malic acid via the Ley's sulfone-based α-amidalkylation protocol (dr = 6.8:1), with the chiral pre-ω-chain. The latter was synthesized in high enantioselectivity from (E)-2-octenol by the Sharpless asymmetric epoxidation and the titanocene-mediated epoxide opening. This modular approach is quite concise and flexible, and requires only eight steps from commercially available reagents.
Sigma-Aldrich
Prostaglandin E1, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E1, ≥98% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E1, powder, γ-irradiated, BioXtra, suitable for cell culture
Alprostadil, European Pharmacopoeia (EP) Reference Standard