Merck
  • Axial interactions in the mixed-valent CuA active site and role of the axial methionine in electron transfer.
Axial interactions in the mixed-valent CuA active site and role of the axial methionine in electron transfer.
Proceedings of the National Academy of Sciences of the United States of America (2013-08-22)
Ming-Li Tsai, Ryan G Hadt, Nicholas M Marshall, Tiffany D Wilson, Yi Lu, Edward I Solomon
ABSTRACT
Within Cu-containing electron transfer active sites, the role of the axial ligand in type 1 sites is well defined, yet its role in the binuclear mixed-valent CuA sites is less clear. Recently, the mutation of the axial Met to Leu in a CuA site engineered into azurin (CuA Az) was found to have a limited effect on E(0) relative to this mutation in blue copper (BC). Detailed low-temperature absorption and magnetic circular dichroism, resonance Raman, and electron paramagnetic resonance studies on CuA Az (WT) and its M123X (X = Q, L, H) axial ligand variants indicated stronger axial ligation in M123L/H. Spectroscopically validated density functional theory calculations show that the smaller ΔE(0) is attributed to H2O coordination to the Cu center in the M123L mutant in CuA but not in the equivalent BC variant. The comparable stabilization energy of the oxidized over the reduced state in CuA and BC (CuA ∼ 180 mV; BC ∼ 250 mV) indicates that the S(Met) influences E(0) similarly in both. Electron delocalization over two Cu centers in CuA was found to minimize the Jahn-Teller distortion induced by the axial Met ligand and lower the inner-sphere reorganization energy. The Cu-S(Met) bond in oxidized CuA is weak (5.2 kcal/mol) but energetically similar to that of BC, which demonstrates that the protein matrix also serves an entatic role in keeping the Met bound to the active site to tune down E(0) while maintaining a low reorganization energy required for rapid electron transfer under physiological conditions.
Sigma-Aldrich
Azurin, from Pseudomonas aeruginosa, lyophilized powder
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
SAFC
L-Methionine
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Methionine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Methionine, Pharmaceutical Secondary Standard; Certified Reference Material