Merck
  • Dynamic regulation of Ero1α and peroxiredoxin 4 localization in the secretory pathway.
Dynamic regulation of Ero1α and peroxiredoxin 4 localization in the secretory pathway.
The Journal of biological chemistry (2013-08-28)
Taichi Kakihana, Kazutaka Araki, Stefano Vavassori, Shun-ichiro Iemura, Margherita Cortini, Claudio Fagioli, Tohru Natsume, Roberto Sitia, Kazuhiro Nagata
ABSTRACT
In the early secretory compartment (ESC), a network of chaperones and enzymes assists oxidative folding of nascent proteins. Ero1 flavoproteins oxidize protein disulfide isomerase (PDI), generating H2O2 as a byproduct. Peroxiredoxin 4 (Prx4) can utilize luminal H2O2 to oxidize PDI, thus favoring oxidative folding while limiting oxidative stress. Interestingly, neither ER oxidase contains known ER retention signal(s), raising the question of how cells prevent their secretion. Here we show that the two proteins share similar intracellular localization mechanisms. Their secretion is prevented by sequential interactions with PDI and ERp44, two resident proteins of the ESC-bearing KDEL-like motifs. PDI binds preferentially Ero1α, whereas ERp44 equally retains Ero1α and Prx4. The different binding properties of Ero1α and Prx4 increase the robustness of ER redox homeostasis.
Sigma-Aldrich
Protein Disulfide Isomerase from bovine liver, ≥100 units/mg protein, lyophilized powder
Pricing and availability is not currently available.