Merck
  • Deacetylase Rpd3 facilitates checkpoint adaptation by preventing Rad53 overactivation.
Deacetylase Rpd3 facilitates checkpoint adaptation by preventing Rad53 overactivation.
Molecular and cellular biology (2013-08-28)
Ran Tao, Hua Xue, Jianping Zhang, Jieyuan Liu, Haiteng Deng, Ye-Guang Chen
ABSTRACT
The DNA damage checkpoint is tightly controlled. After its activation, the checkpoint machinery is inactivated once lesions are repaired or undergoes adaptation if the DNA damage is unable to be repaired. Protein acetylation has been shown to play an important role in DNA damage checkpoint activation. However, the role of acetylation in checkpoint inactivation is unclear. Here we show that histone deacetylase Rpd3-mediated deacetylation of Rad53 plays an important role in checkpoint adaptation. Deletion of Rpd3 or inhibition of its activity impairs adaptation. RPD3 deletion also leads to a higher acetylation level and enhanced kinase activity of Rad53. Replacement of two major acetylation sites of Rad53 with arginine reduces its activity and further suppresses the adaptation defect of rpd3Δ cells, indicating that Rpd3 facilitates adaptation by preventing Rad53 overactivation. Similar to its role in adaptation, deletion of RPD3 or inhibition of its activity also suppressed checkpoint recovery. Altogether, our findings reveal an important role of Rpd3 in promoting checkpoint adaptation via deacetylation and inhibition of Rad53.
Product Description
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propylpentanoic acid
Sigma-Aldrich
Fructose-6-phosphate Kinase from Bacillus stearothermophilus, Type VII, lyophilized powder, ≥50 units/mg protein
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®