Merck
  • A common reference-based indirect comparison meta-analysis of intravenous valproate versus intravenous phenobarbitone for convulsive status epilepticus.
A common reference-based indirect comparison meta-analysis of intravenous valproate versus intravenous phenobarbitone for convulsive status epilepticus.
Epileptic disorders : international epilepsy journal with videotape (2013-08-29)
Francesco Brigo, Stanley C Igwe, Raffaele Nardone, Frediano Tezzon, Luigi Giuseppe Bongiovanni, Eugen Trinka
ABSTRACT
To compare intravenous valproate (IV-VPA) with intravenous phenobarbitone (IV-PB) in the treatment of established generalised convulsive status epilepticus (GCSE). Efficacy and safety were estimated using a common-reference based indirect comparison meta-analysis (CRBMA) methodology. Randomised controlled trials (RCTs) investigating the use of IV-VPA or IV-PB versus intravenous phenytoin (IV-PHT) for GCSE were identified by a systematic search of the literature. A random effects model was used to estimate Mantel-Haenszel odds ratios (ORs) for efficacy and safety of IV-VPA or IV-PB versus IV-PHT in a standard meta-analysis. Adjusted indirect comparisons were then made between VPA and PB using the obtained results. CRBMA showed that VPA does not lead to significantly higher seizure cessation (OR 1.00; 95% CI: 0.36-2.76) compared to PB, although it exhibits fewer adverse effects (OR 0.17; 95% CI: 0.04-0.71). Results of this CRBMA are consistent with results of a recently published head-to-head comparison of IV-VPA and IV-PB. There is insufficient evidence to demonstrate superiority of IV-VPA over IV-PB for the treatment of GCSE in terms of efficacy. Some direct and indirect comparisons suggest that VPA has a better safety profile than PB. However, the limited numbers of underpowered RCTs included in this meta-analysis are not sufficient to justify a change in clinical practice. More rigorous and appropriately powered RCTs are therefore required to definitively determine the efficacy and tolerability of VPA for the treatment of GCSE.
Product Description
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenobarbital solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
2-Propylpentanoic acid
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®