Merck
  • The relation between symptom improvement and gastric emptying in the treatment of diabetic and idiopathic gastroparesis.
The relation between symptom improvement and gastric emptying in the treatment of diabetic and idiopathic gastroparesis.
The American journal of gastroenterology (2013-09-06)
Pieter Janssen, M Scott Harris, Mike Jones, Tatsuhiro Masaoka, Ricard Farré, Hans Törnblom, Lukas Van Oudenhove, Magnus Simrén, Jan Tack
ABSTRACT
The relationship between symptom improvement (SI) and acceleration of gastric emptying (GE) for different drugs used in the treatment of idiopathic and diabetic gastroparesis is uncertain. In this paper we examined the study-specific correlations between SI and GE, and we performed a meta-regression analysis of the association across multiple studies. The MEDLINE database (1,946 to present) was searched, and only controlled trials or trials with an established effective comparator that compared both SI and GE were included. Studies were identified for metoclopramide (n=6), domperidone (n=6), cisapride (n=14), erythromycin (n=3), botulinum toxin (n=2), and levosulpiride (n=3). Even though most drugs concomitantly improved symptoms and accelerated GE, no study reported a significant correlation between SI and GE. Moreover, a correlation analysis over all studies using meta-regression did not show a significant relationship between SI and GE. Our findings need to be qualified by inconsistencies in study methods, which is a limitation but also suggests that our findings are robust to methodological factors. In this review, no evidence of a relationship between SI and GE was identified for different drugs used for the treatment of gastroparesis. This finding questions the use of GE measurement to direct drug development for gastroparesis.
Product Description
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
(±)-Sulpiride
Sigma-Aldrich
(S)-(−)-Sulpiride, ≥98% (titration)
Domperidone, European Pharmacopoeia (EP) Reference Standard
Supelco
Metoclopramide, VETRANAL®, analytical standard
Supelco
(S)-(−)-Sulpiride, VETRANAL®, analytical standard
USP
Metoclopramide hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Domperidone, powder, ≥98% (HPLC)
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.