Merck
Survival of foodborne pathogens on inshell walnuts.
International journal of food microbiology (2013-09-13)
Tyann Blessington, Christopher G Theofel, Elizabeth J Mitcham, Linda J Harris
ABSTRACT
The survival of Salmonella enterica Enteritidis PT 30 or five-strain cocktails of S. enterica, Escherichia coli O157:H7, and Listeria monocytogenes was evaluated on inshell walnuts during storage. Inshell walnuts were separately inoculated with an aqueous preparation of the pathogens at levels of 10 to 4 log CFU/nut, dried for 24 h, and then stored at either 4 °C or ambient conditions (23-25 °C, 25-35% relative humidity) for 3 weeks to more than 1 year. During the initial 24-h drying period, bacterial levels declined by 0.7 to 2.4 log CFU/nut. After the inoculum dried, further declines of approximately 0.1 log CFU/nut per month of Salmonella Enteritidis PT 30 levels were observed on inshell walnuts stored at 4 °C; at ambient conditions the rates of decline ranged from 0.55 to 2.5 log CFU/nut per month. Rates of decline were generally greater during the first few weeks of storage, particularly at lower inoculum levels. The survival of the five-strain cocktails inoculated at very low levels (under 400 CFU/nut) was determined during storage at ambient conditions. The pathogens could be recovered by either enumeration or enrichment from most samples throughout the 3-month storage period; reductions in bacterial levels from the beginning to end of storage were 0.7, 0.2, and 2.3 log CFU/nut for Salmonella, E. coli O157:H7, and L. monocytogenes, respectively. For 6% of all nut samples (14 of 234 samples), pathogens were isolated from the second but not first 24-h enrichment, suggesting that bacterial cells were viable but not easily culturable. Salmonella-inoculated walnuts were exposed for 2 min to water or a 3% solution of sodium hypochlorite (to mimic commercial brightening) either 24 h or 7 days after inoculation; treated nuts were dried for 24h and held at ambient conditions. Salmonella levels were reduced by less than 0.5 log or 2.4 to 2.6 log CFU/nut on water- or chlorine- treated walnuts, respectively, regardless of postinoculation treatment time. Additional reductions of 2.6 and 2.1 log CFU/nut were observed for water- and chlorine-treated walnuts, respectively, after storage for 2 weeks at ambient conditions. Bacterial foodborne pathogens are capable of long-term survival on the surface of inshell walnuts even when initial levels are low.