Merck
  • Chromatographic methods for simultaneous determination of diiodohydroxyquinoline and metronidazole in their binary mixture.
Chromatographic methods for simultaneous determination of diiodohydroxyquinoline and metronidazole in their binary mixture.
Pakistan journal of pharmaceutical sciences (2013-09-17)
Nouruddin Wageih Ali, Mohammed Gamal, Mohammed Abdelkawy
ABSTRACT
Two chromatographic methods were developed for analysis ofdiiodohydroxyquinoline (DIHQ) and metronidazole (MTN). In the first method, diiodohydroxyquinoline and metronidazole were separated on TLC silica gel 60F254 plate using chloroform: acetone: glacial acetic acid (7.5: 2.5: 0.1, by volume) as mobile phase. The obtained bands were then scanned at 254 nm. The second method is a RP-HPLC method in which diiodohydroxyquinoline and metronidazole were separated on a reversed-phase C18 column using water : methanol (60 :40, V/V, PH=3.6 )as mobile phase at a flow rate of 0.7 mL.min-1 and UV detection at 220 nm. The mentioned methods were successfully used for determination of diiodohydroxyquinoline and metronidazole in pure form and in their pharmaceutical formulation.
Product Description
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Metronidazole, BioXtra
Supelco
Metronidazole, VETRANAL®, analytical standard
Supelco
Metronidazole, analytical standard