Merck
  • Analgesia induced by 2- or 100-Hz electroacupuncture in the rat tail-flick test depends on the anterior pretectal nucleus.
Analgesia induced by 2- or 100-Hz electroacupuncture in the rat tail-flick test depends on the anterior pretectal nucleus.
Life sciences (2013-09-26)
Marcelo L Silva, Josie R T Silva, Wiliam A Prado
ABSTRACT
The anterior pretectal nucleus (APtN) and electroacupuncture (EA) activate descending mechanisms to modulate nociceptive inputs in the spinal dorsal horn. This study examines qualitatively whether mechanisms in the APtN participate in the EA-induced analgesia in rats. The tail-flick test was utilized to examine the changes produced by non-selective antagonists of serotonergic (methysergide, 37 pg), muscarinic (atropine, 10 ng) and opioid (naloxone, 10 ng) receptors; selective antagonists against μ (CTOP, 6.4 μg), δ (ICI174,864, 6.9 μg) or κ (nor-BNI, 7.3 μg); 5HT1 (methiothepin, 0.47 μg), 5HT2 (ketanserin, 5.4 μg), or 5HT3 (MDL 72222, 15.7 μg); and GABAA (bicuculline, 150 ng) receptors injected into the dorsal (d) or ventral (v) APtN on the antinociception induced by a 20-min EA applied at 2- or 100-Hz frequency to the Zusanli and Sanyinjiao acupoints. The 2-Hz EA-induced analgesia was blocked by naloxone, CTOP or atropine, was less intense after bicuculline, was shorter after methysergide or methiothepin in dAPtN, and was less intense after methysergide, methiothepin and bicuculline in vAPtN. The 100-Hz EA-induced analgesia was less intense after methysergide, methiothepin and CTOP in vAPtN, and remained unchanged after injection of the antagonists into the dAPtN. The 2-Hz EA-induced analgesia utilizes cholinergic muscarinic, μ-opioid, GABAA and 5-HT1 mechanisms in the dAPtN and μ-opioid and 5-HT1 mechanisms in the vAPtN, while 100-Hz EA-induced analgesia utilizes μ-opioid and 5-HT1 mechanisms in the vAPtN but does not utilize them in the dAPtN.
Sigma-Aldrich
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder
Sigma-Aldrich
CTOP, ≥97% (HPLC)
Sigma-Aldrich
(+)-Bicuculline, ≥97.0% (TLC)
Supelco
Atropine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Naltrexone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Naloxone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®