Merck
  • Importance of polarity of the α4-α5 loop residue-Asn(166) in the pore-forming domain of the Bacillus thuringiensis Cry4Ba toxin: implications for ion permeation and pore opening.
Importance of polarity of the α4-α5 loop residue-Asn(166) in the pore-forming domain of the Bacillus thuringiensis Cry4Ba toxin: implications for ion permeation and pore opening.
Biochimica et biophysica acta (2013-10-15)
Thanate Juntadech, Yodsoi Kanintronkul, Chalermpol Kanchanawarin, Gerd Katzenmeier, Chanan Angsuthanasombat
ABSTRACT
Bacillus thuringiensis Cry4Ba toxin is lethal to mosquito-larvae by forming ion-permeable pores in the target midgut cell membrane. Previously, the polarity of Asn(166) located within the α4-α5 loop composing the Cry4Ba pore-forming domain was shown to be crucial for larvicidal activity. Here, structurally stable-mutant toxins of both larvicidal-active (N166D) and inactive (N166A and N166I) mutants were FPLC-purified and characterized for their relative activities in liposomal-membrane permeation and single-channel formation. Similar to the 65-kDa trypsin-activated wild-type toxin, the N166D bio-active mutant toxin was still capable of releasing entrapped calcein from lipid vesicles. Conversely, the two other bio-inactive mutants showed a dramatic decrease in causing membrane permeation. When the N166D mutant was incorporated into planar lipid bilayers (under symmetrical conditions at 150mM KCl, pH8.5), it produced single-channel currents with a maximum conductance of about 425pS comparable to the wild-type toxin. However, maximum conductances for single K(+)-channels formed by both bio-inactive mutants (N166I and N166A) were reduced to approximately 165-205pS. Structural dynamics of 60-ns simulations of a trimeric α4-α5 pore model in a fully hydrated-DMPC system revealed that an open-pore structure could be observed only for the simulated pores of the wild type and N166D. Additionally, the number of lipid molecules interacting with both wild-type and N166D pores is relatively higher than those of N166A and N166I pores. Altogether, our results further signify that the polarity at the α4-α5 loop residue-Asn(166) is directly involved in ion permeation through the Cry4Ba toxin-induced ionic pore and pore opening at the membrane-water interface.
Product Description
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Alanine, ≥99%
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
SAFC
L-Isoleucine
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
L-Asparagine, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Asparagine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Asparagine, BioReagent, suitable for cell culture, suitable for insect cell culture