Merck
  • Activity of chalcones derived from 2,4,5-trimethoxybenzaldehyde against Meloidogyne exigua and in silico interaction of one chalcone with a putative caffeic acid 3-O-methyltransferase from Meloidogyne incognita.
Activity of chalcones derived from 2,4,5-trimethoxybenzaldehyde against Meloidogyne exigua and in silico interaction of one chalcone with a putative caffeic acid 3-O-methyltransferase from Meloidogyne incognita.
Experimental parasitology (2013-10-22)
Alexandro Silva Nunes, Vicente Paulo Campos, Alessandra Mascarello, Taisa Regina Stumpf, Louise Domenghini Chiaradia-Delatorre, Alan Rodrigues Teixeira Machado, Helvécio Martins Santos Júnior, Rosendo Augusto Yunes, Ricardo José Nunes, Denilson Ferreira Oliveira
ABSTRACT
Meloidogyne exigua is a parasitic nematode of plants that causes great losses to coffee farmers. In an effort to develop parasitic controls, 154 chalcones were synthesized and screened for activity against this nematode. The best results were obtained with (2E)-1-(4'-nitrophenyl)-3-(2,4,5-trimethoxyphenyl)prop-2-en-1-one (6) with a 50% lethal concentration (LC50) of 171 μg/ml against M. exigua second-stage juveniles, in comparison to the commercially-available nematicide carbofuran which had an LC50 of 260 μg/ml under the same conditions. When coffee plants were used, 6 reduced the nematode population to ~50% of that observed in control plants. To investigate the mechanism of action of 6, an in silico study was carried out, which indicated that 6 may act against M. exigua through inhibition of a putative caffeic acid 3-O-methyltransferase homodimer, the amino acid sequence of which was determined by examining the genome of Meloidogyne incognita.