Merck
  • Cholera toxin breakdowns oral tolerance via activation of canonical NF-κB.
Cholera toxin breakdowns oral tolerance via activation of canonical NF-κB.
Cellular immunology (2013-10-22)
Kyoung-Jin Kim, Han-A Kim, Kook Heon Seo, Hern-Ku Lee, Bok Yun Kang, Suhn-Young Im
ABSTRACT
The mechanisms of mucosal immunogenicity and adjuvanticity of bacterial exotoxins remains unknown. In this study, we investigated the role of the transcription factor nuclear factor-κB (NF-κB) in cholera toxin (CT)-induced alteration of oral tolerance. Feeding CT abrogated ovalbumin (OVA)-induced oral tolerance, as evaluated by OVA-specific serum antibody responses, and CD4(+) T cell proliferation. CT feeding activated canonical NF-κB (one heterodimer type, p50-p65) and mRNA expression of NF-κB-dependent proinflammatory cytokines in mesenteric lymph node (MLN) and Peyer's patch (PP) cells. CT no longer showed abrogation of oral tolerance in mice pretreated with p50 small interfering RNAs (siRNAs). ADP-ribosylation inhibitors inhibited CT-induced NF-κB activation. These data suggest that CT induces canonical NF-κB activation in intestinal lymphoid cells, which plays a key role in mucosal immunogenicity and adjuvanticity.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Albumin from chicken egg white, powder, 62-88% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥90% (agarose gel electrophoresis)
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)