Merck
  • Decoding the histone code: Role of H3K36me3 in mismatch repair and implications for cancer susceptibility and therapy.
Decoding the histone code: Role of H3K36me3 in mismatch repair and implications for cancer susceptibility and therapy.
Cancer research (2013-10-23)
Guo-Min Li
ABSTRACT
DNA mismatch repair (MMR) maintains genome stability primarily by correcting replication-associated mismatches. Defects in MMR lead to several human cancers characterized by frequent alterations in simple repetitive DNA sequences, a phenomenon called microsatellite instability (MSI). In most MSI-positive cancers, genetic or epigenetic changes that alter the function or expression of an essential MMR protein have been identified. However, in a subset of MSI-positive cancers, epigenetic or genetic changes have not been found in known MMR genes, such that the molecular basis of the MMR defect in these cells remains unknown. A possible answer to this puzzle emerged recently when it was discovered that H3K36me3, a well-studied posttranslational histone modification or histone mark, plays a role in regulating human MMR in vivo. In this review, potential roles for this histone mark to modulate genome stability and cancer susceptibility in human cells are discussed.
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%