Merck
  • Interactions between lens epithelial and fiber cells reveal an intrinsic self-assembly mechanism.
Interactions between lens epithelial and fiber cells reveal an intrinsic self-assembly mechanism.
Developmental biology (2013-11-12)
L J Dawes, Y Sugiyama, F J Lovicu, C G Harris, E J Shelley, J W McAvoy
ABSTRACT
How tissues and organs develop and maintain their characteristic three-dimensional cellular architecture is often a poorly understood part of their developmental program; yet, as is clearly the case for the eye lens, precise regulation of these features can be critical for function. During lens morphogenesis cells become organized into a polarized, spheroidal structure with a monolayer of epithelial cells overlying the apical tips of elongated fiber cells. Epithelial cells proliferate and progeny that shift below the lens equator differentiate into new fibers that are progressively added to the fiber mass. It is now known that FGF induces epithelial to fiber differentiation; however, it is not fully understood how these two forms of cells assemble into their characteristic polarized arrangement. Here we show that in FGF-treated epithelial explants, elongating fibers become polarized/oriented towards islands of epithelial cells and mimic their polarized arrangement in vivo. Epithelial explants secrete Wnt5 into the culture medium and we show that Wnt5 can promote directed behavior of lens cells. We also show that these explants replicate aspects of the Notch/Jagged signaling activity that has been shown to regulate proliferation of epithelial cells in vivo. Thus, our in vitro study identifies a novel mechanism, intrinsic to the two forms of lens cells, that facilitates self-assembly into the polarized arrangement characteristic of the lens in vivo. In this way the lens, with its relatively simple cellular composition, serves as a useful model to highlight the importance of such intrinsic self-assembly mechanisms in tissue developmental and regenerative processes.
Sigma-Aldrich
Fibroblast Growth Factor from bovine pituitary, suitable for cell culture