Merck
  • Transverse spin Seebeck effect versus anomalous and planar Nernst effects in Permalloy thin films.
Transverse spin Seebeck effect versus anomalous and planar Nernst effects in Permalloy thin films.
Physical review letters (2013-11-19)
M Schmid, S Srichandan, D Meier, T Kuschel, J-M Schmalhorst, M Vogel, G Reiss, C Strunk, C H Back
ABSTRACT
Transverse magnetothermoelectric effects are studied in Permalloy thin films grown on MgO and GaAs substrates and compared to those grown on suspended SiN(x) membranes. The transverse voltage along platinum strips patterned on top of the Permalloy films is measured versus the external magnetic field as a function of the angle and temperature gradients. After the identification of the contribution of the planar and anomalous Nernst effects, we find an upper limit for the transverse spin Seebeck effect, which is several orders of magnitude smaller than previously reported.
Sigma-Aldrich
Gallium, 99.9995% trace metals basis
Sigma-Aldrich
Gallium, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Gallium arsenide, (single crystal substrate), <100>, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Magnesium oxide, light, 95%
Sigma-Aldrich
Gallium, 99.999% trace metals basis
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Magnesium oxide, ACS reagent, 97%
Sigma-Aldrich
Magnesium oxide, -10-+50 mesh, 98%
Sigma-Aldrich
Magnesium oxide, ≥99% trace metals basis, -325 mesh