Merck
  • Synthesis of mesoporous SAPO-34 zeolite from mesoporous silica materials for methanol to light olefins.
Synthesis of mesoporous SAPO-34 zeolite from mesoporous silica materials for methanol to light olefins.
Journal of nanoscience and nanotechnology (2013-11-20)
Eun A Kang, Tae-Wan Kim, Ho-Jeong Chae, Min Kim, Kwang-Eun Jeong, Joo-Wan Kim, Chul-Ung Kim, Soon-Yong Jeong
ABSTRACT
Mesoporous SAPO-34 zeolites were synthesized by using as-prepared mesoporous silica material as both silica source and mesopore tailor. The mesoporous SAPO-34 zeolite materials thus obtained are characterized by a series of different techniques, including poweder X-ray diffraction pattern, nitrogen physisorption analysis, scanning electron micrograph, temperature programmed desorption of ammonia, and inductively coupled plasma atomic emission spectrometry. The resultant mesoporous SAPO-34 crystals exhibit sphere-like particle with zeolite layer units. The mesopore size distribution and particle size can be changed by amounts of silica source and water. The methanol-to-olefins (MTO) reactions using these mesoporous SAPO-34 zeolites are carried out with a fixed-bed reactor. Catalytic tests exhibit that the mesoporous SAPO-34 zeolite materials show high catalytic activity compared with the conventional SAPO-34 for MTO reaction. The better catalytic activity and longer life time of the mesoporous SAPO-34 catalysts in MTO are mainly due to the existence of the mesoporosity of SAPO-34 with small particle size.
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, ≥99.9% trace metals basis
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O