Merck
  • Total synthesis of human galanin-like peptide through an aspartic acid ligation.
Total synthesis of human galanin-like peptide through an aspartic acid ligation.
Organic letters (2013-11-26)
Xiaoyang Guan, Matthew R Drake, Zhongping Tan
ABSTRACT
Human galanin-like peptide (hGALP) is a newly discovered hypothalamic peptide that plays important roles in the regulation of food intake and energy balance. Here, we demonstrate that the aspartic acid ligation can be employed to achieve an efficient synthesis of hGALP. The total synthesis of hGALP enhances our ability to study its biology and facilitates the development of more stable analogues.
Product Description
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)