Merck
  • A geographical discrimination of Shanxi extra aged vinegars using polyalcohols as the discriminators.
A geographical discrimination of Shanxi extra aged vinegars using polyalcohols as the discriminators.
Journal of AOAC International (2013-11-29)
Meizhong Luo, Yanjie Zheng, Cen Xiong, Bifang Li, Sujuan Chen, Wenliang Bai, Yongting Zeng, Yongle Li, Xieguang Zhang
ABSTRACT
A discrimination method based on polyalcohol determination was developed for authenticity of protected geographical indication (PGI) vinegars-Shanxi extra aged vinegar (SVs) in China. Six polyalcohols in vinegars including erythritol, arabitol, xylitol, inositol, mannitol, and sorbitol were selected as the PGI discriminators. GC/MS was used to analyze the polyalcohols in the SVs, Zhenjiang vinegars (ZVs), Kazuo aged vinegars (KVs), and other non-geographical indication protected vinegars (NVs). SVs can be distinguished from KVs by the chemical markers mannitol and sorbitol, although the production processes for both types of vinegars are similar. Principal component analysis (PCA) was used to distinguish SVs from ZVs and NVs. The differences among the three kinds of vinegars shown by PCA results may be due to the higher erythritol content in SVs, and the inositol and mannitol in ZVs. This study also found that the amount of polyalcohols in Chinese vinegars increases with the acidity value only, regardless of the aging time. The overall results indicated that the polyalcohols can be practicable discriminators for SV discrimination.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
Xylitol, ≥99% (GC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), Molecular Biology
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Xylitol