Merck
  • Cobalt precursors for high-throughput discovery of base metal asymmetric alkene hydrogenation catalysts.
Cobalt precursors for high-throughput discovery of base metal asymmetric alkene hydrogenation catalysts.
Science (New York, N.Y.) (2013-11-30)
Max R Friedfeld, Michael Shevlin, Jordan M Hoyt, Shane W Krska, Matthew T Tudge, Paul J Chirik
ABSTRACT
Asymmetric hydrogenation of alkenes is one of the most widely used methods for the preparation of single enantiomer compounds, especially in the pharmaceutical and agrochemical industries. For more than four decades, precious metal complexes containing rhodium, iridium, and ruthenium have been predominantly used as catalysts. Here, we report rapid evaluation of libraries of chiral phosphine ligands with a set of simple cobalt precursors. From these studies, base metal precatalysts have been discovered for the hydrogenation of functionalized and unfunctionalized olefins with high enantiomeric excesses, demonstrating the potential utility of more earth-abundant metals in asymmetric hydrogenation.
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis