Merck
  • Femtosecond dynamics of short-range protein electron transfer in flavodoxin.
Femtosecond dynamics of short-range protein electron transfer in flavodoxin.
Biochemistry (2013-12-03)
Ting-Fang He, Lijun Guo, Xunmin Guo, Chih-Wei Chang, Lijuan Wang, Dongping Zhong
ABSTRACT
Intraprotein electron transfer (ET) in flavoproteins is important for understanding the correlation of their redox, configuration, and reactivity at the active site. Here, we used oxidized flavodoxin as a model system and report our complete characterization of a photoinduced redox cycle from the initial charge separation in 135-340 fs to subsequent charge recombination in 0.95-1.6 ps and to the final cooling relaxation of the product(s) in 2.5-4.3 ps. With 11 mutations at the active site, we observed that these ultrafast ET dynamics, much faster than active-site relaxation, mainly depend on the reduction potentials of the electron donors with minor changes caused by mutations, reflecting a highly localized ET reaction between the stacked donor and acceptor at a van der Waals distance and leading to a gas-phase type of bimolecular ET reaction confined in the active-site nanospace. Significantly, these ultrafast ET reactions ensure our direct observation of vibrationally excited reaction product(s), suggesting that the back ET barrier is effectively reduced because of the decrease in the total free energy in the Marcus inverted region, leading to the accelerated charge recombination. Such vibrationally coupled charge recombination should be a general feature of flavoproteins with similar configurations and interactions between the cofactor flavin and neighboring aromatic residues.
Sigma-Aldrich
Riboflavin Sodium Phosphate Hydrate, tested according to Ph. Eur.
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
SAFC
L-Tyrosine
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, 70-79% (fluorimetric)
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, synthetic, ≥70% (HPLC)
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, FG
Riboflavin sodium phosphate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%