Merck
Earliest evidence for commensal processes of cat domestication.
Proceedings of the National Academy of Sciences of the United States of America (2013-12-18)
Yaowu Hu, Songmei Hu, Weilin Wang, Xiaohong Wu, Fiona B Marshall, Xianglong Chen, Liangliang Hou, Changsui Wang
ABSTRACT
Domestic cats are one of the most popular pets globally, but the process of their domestication is not well understood. Near Eastern wildcats are thought to have been attracted to food sources in early agricultural settlements, following a commensal pathway to domestication. Early evidence for close human-cat relationships comes from a wildcat interred near a human on Cyprus ca. 9,500 y ago, but the earliest domestic cats are known only from Egyptian art dating to 4,000 y ago. Evidence is lacking from the key period of cat domestication 9,500-4,000 y ago. We report on the presence of cats directly dated between 5560-5280 cal B.P. in the early agricultural village of Quanhucun in Shaanxi, China. These cats were outside the wild range of Near Eastern wildcats and biometrically smaller, but within the size-range of domestic cats. The δ(13)C and δ(15)N values of human and animal bone collagen revealed substantial consumption of millet-based foods by humans, rodents, and cats. Ceramic storage containers designed to exclude rodents indicated a threat to stored grain in Yangshao villages. Taken together, isotopic and archaeological data demonstrate that cats were advantageous for ancient farmers. Isotopic data also show that one cat ate less meat and consumed more millet-based foods than expected, indicating that it scavenged among or was fed by people. This study offers fresh perspectives on cat domestication, providing the earliest known evidence for commensal relationships between people and cats.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture