Merck
  • Clinical and radiographic evaluation of single-tooth dental implants placed in grafted extraction sites: a one-year report.
Clinical and radiographic evaluation of single-tooth dental implants placed in grafted extraction sites: a one-year report.
Journal of the International Academy of Periodontology (2013-12-25)
Ahmad Kutkut, Sebastiano Andreana, Edward Monaco
ABSTRACT
The aim of this report was to clinically and radiographically evaluate changes to the hard and soft tissues around implants placed in extraction sockets grafted with medical grade calcium sulfate hemihydrate (MGCSH) mixed with platelet-rich plasma (PRP) and a collagen resorbable plug after one year of function. This evaluation was part of a previous study conducted to evaluate extraction socket grafts. Fourteen subjects out of 16 were evaluated. After tooth extraction, eight subjects received MGCSH mixed with PRP in the extraction sockets (test group), and six subjects received collagen resorbable plug dressing material (control group). After three months of bone healing, dental implants were placed. Three months after implant installation, provisional restorations were placed and implants were loaded in function for one month followed by definitive restorations. Follow-up examinations and intraoral digital radiographs were made at baseline and one year after definitive restorations to evaluate the marginal bone level in each subject. At the one-year follow-up, the survival and success rate was 100% for all implants. There was no statistically significant difference in the amount of vertical bone loss between groups after 1 year (p > 0.05). For the test group, there was a mean mesial bone loss of -0.8 +/- 0.6mm and a mean distal bone loss of -0.5 +/- 0.4mm. For the control group, there was a mean mesial bone loss of -1.1 +/- 0.7mm and a mean distal bone loss of -0.6 +/- 0.6mm. At the one-year follow-up, the implant placement in grafted sockets was not affected by the type of the graft material. Implants placed in sockets grafted with MGCSH mixed with PRP showed less marginal bone loss after one year in comparison to those with collagen resorbable grafts.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%
Sigma-Aldrich
Calcium sulfate dihydrate, ACS reagent, 98%
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Sigma-Aldrich
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Sigma-Aldrich
Calcium sulfate dihydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Sigma-Aldrich
Drierite, with indicator, 6 mesh
Sigma-Aldrich
Drierite, with indicator, 4 mesh
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh