Merck
  • Methylene blue used in the treatment of refractory shock resulting from drug poisoning.
Methylene blue used in the treatment of refractory shock resulting from drug poisoning.
Clinical toxicology (Philadelphia, Pa.) (2013-12-25)
J Fisher, G Taori, G Braitberg, A Graudins
ABSTRACT
Methylene blue inhibits the nitric oxide-cyclic guanosine monophosphate (NO-cGMP) pathway, decreasing vasodilation and increasing responsiveness to vasopressors. It is reported to improve haemodynamics in distributive shock from various causes including septicaemia and post-cardiac surgery. Reports of use in overdose are limited. We describe the use of methylene blue to treat a case of refractory distributive shock following a mixed drug poisoning. A 41-year-old male presented following reported ingestion of 18 g extended-release quetiapine, 10 g controlled-release carbamazepine, 240 mg fluoxetine, 35 g enteric-coated sodium valproate and 375 mg oxazepam. He was comatose and intubated on presentation. Progressive hypotension developed. Echocardiogram revealed a hyperdynamic left ventricle, suggesting distributive shock. The patient remained hypotensive despite intravenous fluid boluses, escalating vasopressor infusions. Arterial blood gas revealed metabolic acidaemia and high lactate. Methylene blue was administered as loading-dose of 1.5 mg/kg and continuous infusion (1.5 mg/kg/h for 12 h, then 0.75 mg/kg/h for 12 h) resulting in rapid improvement in haemodynamic parameters and weaning of vasopressors. Serum quetiapine concentration was 18600 ng/mL (30-160 ng/mL), collected at the time of peak toxicity. Severe quetiapine poisoning produces hypotension primarily from alpha-adrenoreceptor antagonism. Methylene blue may have utility in the treatment of distributive shock resulting from poisoning refractory to standard vasopressor therapy.
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Oxazepam solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Oxazepam for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propylpentanoic acid
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications