Merck
  • Arbidol exhibits strong inhibition towards UDP-glucuronosyltransferase (UGT) 1A9 and 2B7.
Arbidol exhibits strong inhibition towards UDP-glucuronosyltransferase (UGT) 1A9 and 2B7.
Die Pharmazie (2014-01-10)
Xin Liu, Ting Huang, Jian-Xing Chen, Jia Zeng, Xu-Ran Fan, Xu-Zhu, Zhen-Wen Yu, Xiao-Yu Sun, Mo Hong, Hong-Zhi Sun
ABSTRACT
The aim of the present study was to investigate arbidol's inhibition towards UDP-glucuronosyltransferase (UGT) 1A9 and 2B7. The nonspecific probe substrate 4-methylumbelliferone (4-MU) and recombinant UGT enzymes (UGT1A9, UGT2B7) were firstly used to evaluate the inhibition of arbidol towards UGT1A9 and UGT2B7. Furthermore, specific substrates of UGT1A9 and UGT2B7 propofol and zidovudine (AZT) were used to determine the inhibition of arbidol towards UGT1A9 and UGT2B7. Inhibition type and inhibition kinetic parameters (Ki) were determined. In vitro-in vivo extrapolation (IV-IVE) was performed to predict in vivo DDI magnitude induced by arbidol. Arbidol was demonstrated to exhibit competitive inhibition towards UGT1A9 and UGT2B7 without substate-dependent behaviour. The inhibition kinetic parameters (Ki) were calculated to be 0.5 microM, 3.5 microM, 2.8 microM, 29.7 microM for UGT2B7-mediated 4-MU glucuronidation, UGT1A9-mediated 4-MU glucuronidation, UGT2B7-mediated AZT glucuronidation, and UGT1A9-mediated propofol glucuronidation, respectively. Using these parameters, the in vivo alteration of area under of concentration-time curve (AUC) was calculated to be 156%, 22%, 28% and 2.6%, respectively. Given that arbidol exhibits strong inhibition towards UGT1A9 and UGT2B7, clinical monitoring should be given when arbidol was co-administered with drugs mainly undergoing UGT1A9, UGT2B7-mediated metabolism.
Product Description
Propofol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Arbidol hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Sigma-Aldrich
3′-Azido-3′-deoxythymidine, ≥98% (HPLC)
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE
Propofol for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Zidovudine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Propofol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®