Merck
  • Impact of fluorescein on the antimicrobial efficacy of photoactivated riboflavin in corneal collagen cross-linking.
Impact of fluorescein on the antimicrobial efficacy of photoactivated riboflavin in corneal collagen cross-linking.
Journal of refractive surgery (Thorofare, N.J. : 1995) (2014-01-10)
Olivier Richoz, Zisis Gatzioufas, Patrice Francois, Jacques Schrenzel, Farhad Hafezi
ABSTRACT
To investigate the effect of fluorescein on the antimicrobial efficacy of photoactivated riboflavin in corneal collagen cross-linking (CXL) in vitro. The ultraviolet light-A (UVA) absorption by fluorescein and riboflavin in different concentrations was analyzed with a spectrophotometer. The killing rate of Staphylococcus aureus strains after CXL with UV-A irradiation using different solutions containing riboflavin and/or fluorescein, was evaluated in vitro. Fluorescein absorbed UV-A to a significant extent, which augmented with increasing concentration. Moreover, addition of fluorescein to the riboflavin solution contributed to a significant reduction of the killing rate of S. aureus strains after 30 minutes of UV-A irradiation in vitro with a killing rate of 85%, whereas it was 47% in the presence of 2% fluorescein (P = .0247). The results indicate that fluorescein competes with riboflavin for the absorption of UV-A during CXL and reduces the antimicrobial effect of the procedure. The authors recommend that physicians treating infectious ulcerative keratitis by CXL should not stain the cornea with fluorescein for visualization of the corneal ulceration prior to CXL
Product Description
Riboflavin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Supelco
Riboflavin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(−)-Riboflavin, meets USP testing specifications
Sigma-Aldrich
(−)-Riboflavin, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
(−)-Riboflavin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Supelco
Riboflavin (B2), analytical standard
Sigma-Aldrich
Fluorescein sodium salt, p.a.
Sigma-Aldrich
Fluorescein sodium salt, BioReagent, suitable for fluorescence
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Fluorescein sodium salt, used as fluorescent tracer
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance