Merck
  • Pharmacological correction of gating defects in the voltage-gated Ca(v)2.1 Ca²⁺ channel due to a familial hemiplegic migraine mutation.
Pharmacological correction of gating defects in the voltage-gated Ca(v)2.1 Ca²⁺ channel due to a familial hemiplegic migraine mutation.
Neuron (2014-01-15)
Akira Inagaki, C Andrew Frank, Yuriy M Usachev, Morris Benveniste, Amy Lee
ABSTRACT
Voltage-gated ion channels exhibit complex properties, which can be targeted in pharmacological therapies for disease. Here, we report that the pro-oxidant, tert-butyl dihydroquinone (BHQ), modulates Ca(v)2.1 Ca²⁺ channels in ways that oppose defects in channel gating and synaptic transmission resulting from a familial hemiplegic migraine mutation (S218L). BHQ slows deactivation, inhibits voltage-dependent activation, and potentiates Ca²⁺-dependent facilitation of Ca(v)2.1 channels in transfected HEK293T cells. These actions of BHQ help offset the gain of function and reduced Ca²⁺-dependent facilitation of Ca(v)2.1 channels with the S218L mutation. Transgenic expression of the mutant channels at the Drosophila neuromuscular junction causes abnormally elevated evoked postsynaptic potentials and impaired synaptic plasticity, which are largely restored to the wild-type phenotypes by BHQ. Our results reveal a mechanism by which a Ca(v)2.1 gating modifier can ameliorate defects associated with a disease-causing mutation in Ca(v)2.1.
Product Description
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)