Merck
  • Comprehensive genomic profiling of relapsed and metastatic adenoid cystic carcinomas by next-generation sequencing reveals potential new routes to targeted therapies.
Comprehensive genomic profiling of relapsed and metastatic adenoid cystic carcinomas by next-generation sequencing reveals potential new routes to targeted therapies.
The American journal of surgical pathology (2014-01-15)
Jeffrey S Ross, Kai Wang, Janna V Rand, Christine E Sheehan, Timothy A Jennings, Rami N Al-Rohil, Geoff A Otto, John C Curran, Gary Palmer, Sean R Downing, Roman Yelensky, Doron Lipson, Sohail Balasubramanian, Lazaro Garcia, Kristen Mahoney, Siraj M Ali, Vincent A Miller, Philip J Stephens
ABSTRACT
We hypothesized that next-generation sequencing could reveal actionable genomic alterations (GAs) and potentially expand treatment options for patients with advanced adenoid cystic carcinoma (ACC). Genomic profiling using next-generation sequencing was performed on hybridization-captured, adapter ligation libraries derived from 28 relapsed and metastatic formalin-fixed paraffin-embedded ACC. The 3230 exons of 182 cancer-related genes and 37 introns of 14 genes frequently rearranged in cancer were fully sequenced using the Illumina HiSeq 2000. All classes of GAs were evaluated. Actionable GAs were defined as those impacting targeted anticancer therapies on the market or in registered clinical trials. A total of 44 GAs were identified in the 28 ACC tumors, with 12 of 28 (42.9%) of tumors harboring at least 1 potentially actionable GA. The most common nonactionable GAs were identified in KD6MA (5 cases; 18%), ARID1A (4 cases; 14%), RUNX1 (2 cases; 7%), and MYC (2 cases; 7%). Actionable GAs included NOTCH1 (3 cases; 11%), MDM2 (2 cases; 7%), PDGFRA (2 cases; 7%), and CDKN2A/B (p16) (2 cases; 7%). Other potentially actionable GAs identified in a single case included: mutations in AKT1, BAP1, EGFR, and PIK3CA, homozygous deletion of FBXW7, and amplifications of CDK4, FGFR1, IGF1R, KDR, KIT, and MCL1. The frequency of GA in ACC is lower than that seen in the more common solid tumors. Comprehensive genomic profiling of ACC can identify actionable GAs in a subset of patients that could influence therapy for these difficult-to-treat progressive neoplasms.
Product Description
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)