Merck
  • Flexural strength,water sorption and solubility of a methylmethacrylate-free denture base polymer reinforced with glass fibre reinforcement.
Flexural strength,water sorption and solubility of a methylmethacrylate-free denture base polymer reinforced with glass fibre reinforcement.
The European journal of prosthodontics and restorative dentistry (2014-02-01)
M M Mutluay, A Tezvergil-Mutluay, P Vallittu, L Lassila
ABSTRACT
A methylmethacrylate-free denture base polymer (Eclipse) in comparison to a conventional denture base polymer (Palapress vario) was evaluated after water saturation and Stick glass fibre reinforcement. The data were analysed with ANOVA at a = 0.05. Water-storage caused a decrease in the flexural strength and stiffness of the materials (p > 0.05). Conventional denture base material with fibre reinforcement gave highest flexural strength (201.1 MPa) compared to fibre reinforced Eclipse (79.1 MPa) (p < 0.05). Water sorption after 76 days was 2.08% (Palapress vario) and 1.55% (Eclipse). Fibre-reinforcement of methylmethacrylate-free material was not as successful as conventional denture base and needs to be further optimized.
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 20,000
Supelco
Poly(methyl methacrylate), analytical standard, 2,480,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 8,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 10,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 50,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 4,000