Merck
  • Synthesis and enhanced visible-light responsive of C,N,S-tridoped TiO2 hollow spheres.
Synthesis and enhanced visible-light responsive of C,N,S-tridoped TiO2 hollow spheres.
Journal of environmental sciences (China) (2014-02-06)
Xiaoxia Lin, Degang Fu, Lingyun Hao, Zhen Ding
ABSTRACT
C,N,S-tridoped TiO2 hollow spheres (labeled as C,N,S-THs) were synthesized using carbon spheres as template and C,N,S-tridoped TiO2 nanoparticles as building blocks. The structure and physicochemical properties of the catalysts were characterized by Xray diffraction (XRD), scanning electron microscopy (SEM), UV-Vis diffuse reflectance spectrum (DRS), N2 adsorption-desorption isotherms, X-ray photoelectron spectroscopy (XPS) and Photoluminescence emission spectroscopy (PL). The results showed that the hollow spheres had average diameter of about 200 nm and the shell thickness was about 20 nm. The tridoped TiO2 hollow spheres exhibited strong absorption in the visible-light region. C,N,S-tridoped could narrow the band gap of the THs by mixing the orbit O 2p with C 2p, N 2p and S 3p orbits and shift its optical response from ultraviolet (UV) to the visible-light region. PL analysis indicated that the electron-hole recombination rate of TiO2 hollow spheres had been effectively inhibited when doped with C, N and S elements. The photocatalytic activities of the samples were evaluated for the degradation of X-3B (Reactive Brilliant Red dye, C.I. Reactive Red 2) aqueous solution under visible-light (lambda > 420 nm) irradiation. It was found that the C,N,S-tridoped TiO2 hollow spheres indicated higher photocatalytic activity than commercial P25 and the undoped counterpart photocatalyst.
Product Description
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Titanium, ≤10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
sulfur powder, 99.998% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Titanium, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis