Merck
  • Preparation, characterization and application of thiosemicarbazide grafted multiwalled carbon nanotubes for solid-phase extraction of Cd(II), Cu(II) and Pb(II) in environmental samples.
Preparation, characterization and application of thiosemicarbazide grafted multiwalled carbon nanotubes for solid-phase extraction of Cd(II), Cu(II) and Pb(II) in environmental samples.
Journal of environmental sciences (China) (2014-02-21)
Jie Zhang
ABSTRACT
Thiosemicarbazide-grafted multi-walled carbon nanotubes were prepared and employed to investigate the pre-concentration of Cd(II), Cu(II) and Pb(II) from aqueous solution prior to their determination by ICP-OES. The resulting material was characterized by FT-IR, TGA and SEM. Various factors influencing the separation and pre-concentration were investigated. The enrichment factor typically is 60. Under optimized experimental conditions, the maximum adsorption capacities of Cd(II), Cu(II) and Pb(II) were found to be 1.98, 10.94, 3.69 mg/g, and the relative standard deviations are < 3.5% (n = 6). The new adsorbent shows superior reusability and stability. The procedure was successfully applied to the determination of trace quantities of Cd(II), Cu(II) and Pb(II) in water samples.
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, nanopowder, 60-80 nm particle size (APS), ≥99.5% trace metals basis
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Copper, shot, −3-+14 mesh, 99%
Sigma-Aldrich
Thiosemicarbazide, 99%
Sigma-Aldrich
Thiosemicarbazide, puriss. p.a., 98%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Copper, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
Copper, IRMM®, certified reference material, 0.1 mm foil
Copper (O), BCR®, certified reference material, rod
Copper (S, P), BCR®, certified reference material, chips
Copper, IRMM®, certified reference material, 0.5 mm wire