Merck
  • Synthesis and structure-activity relationships of a series of 3-aryl-4-isoxazolecarboxamides as a new class of TGR5 agonists.
Synthesis and structure-activity relationships of a series of 3-aryl-4-isoxazolecarboxamides as a new class of TGR5 agonists.
Bioorganic & medicinal chemistry letters (2010-01-26)
Brian W Budzik, Karen A Evans, David D Wisnoski, Jian Jin, Ralph A Rivero, George R Szewczyk, Channa Jayawickreme, David L Moncol, Hongshi Yu
ABSTRACT
A series of 3-aryl-4-isoxazolecarboxamides identified from a high-throughput screening campaign as novel, potent agonists of the human TGR5 G-protein-coupled receptor is described. Many analogues were readily accessible via solution-phase synthesis which resulted in the rapid identification of key structure-activity relationships (SAR), and the discovery of potent exemplars (up to pEC50=9). Details of the SAR and optimization of this series are presented herein.