Merck
  • Synthesis, structure, and reactivity of low-coordinate 1,1,3,3-tetraethylguanidinate complexes.
Synthesis, structure, and reactivity of low-coordinate 1,1,3,3-tetraethylguanidinate complexes.
Inorganic chemistry (2009-07-25)
Scott D Bunge, Jeffrey A Bertke, Travis L Cleland
ABSTRACT
Diethylcyanamide is added to a hexanes solution of lithium diethylamide [LiN(CH(2)CH(3))(2)] resulting in the formation of lithium 1,1,3,3-tetraethylguanidinate, [Li(mu-TEG)](6) (1). Upon successful isolation of 1, the metathesis reaction of MX(2) (MX(2) = MnBr(2), FeBr(2), CoBr(2), and ZnCl(2)) with [Li(mu-TEG)](6) and lithium bistrimethylsilylamide, LiN(SiMe(3))(2), was performed to generate dinuclear tetraethylguanidinate (TEG) complexes with the general formula [M(mu-TEG){N(SiMe(3))(2)}](2) {M = Mn (2), Fe (3), Co (4), Zn (5)}. Further reaction of 2 with 2 equiv of ethanol (EtOH) and 2 equiv of 2,6-ditert-butylphenol (H-DBP) results in the formation of the manganese alkoxide, [Mn(mu-OEt)(DBP)(H-TEG)](2) (6). Elemental analysis, FT-IR spectroscopy, UV-vis spectroscopy, and single crystal X-ray diffraction were utilized to characterize the six compounds.
Sigma-Aldrich
Diethylcyanamide, 99%
Pricing and availability is not currently available.