Merck
  • Fractionation of Lactoperoxidase and Lactoferrin from Bovine Whey Using a Cation Exchange Membrane
Fractionation of Lactoperoxidase and Lactoferrin from Bovine Whey Using a Cation Exchange Membrane
Journal of Food Science (1997)
Clovis k. Chiu and Mark R.