Merck
  • Studies on the binding site of the galactose-specific agglutinin PA-IL from Pseudomonas aeruginosa.
Studies on the binding site of the galactose-specific agglutinin PA-IL from Pseudomonas aeruginosa.
Glycobiology (1998-05-28)
C P Chen, S C Song, N Gilboa-Garber, K S Chang, A M Wu
ABSTRACT
The binding properties of Pseudomonas aeruginosa agglutinin-I (PA-IL) with glycoproteins (gps) and polysaccharides were studied by both the biotin/avidin-mediated microtiter plate lectin-binding assay and the inhibition of agglutinin-glycan interaction with sugar ligands. Among 36 glycans tested for binding, PA-IL reacted best with two glycoproteins containing Galalpha1-->4Gal determinants and a human blood group ABO precursor equivalent gp, but this lectin reacted weakly or not at all with A and H active gps or sialylated gps. Among the mammalian disaccharides tested by the inhibition assay, the human blood group Pkactive Galalpha1-->4Gal, was the best. It was 7.4-fold less active than melibiose (Galalpha1-->6Glc). PA-IL has a preference for the alpha-anomer in decreasing order as follows: Galalpha1-->6 >Galalpha1-->4 >Galalpha1-->3. Of the monosaccharides studied, the phenylbeta derivatives of Gal were much better inhibitors than the methylbeta derivative, while only an insignificant difference was found between the Galalpha anomer of methyl- and p -NO2-phenyl derivatives. From these results, it can be concluded that the combining size of the agglutinin is as large as a disaccharide of the alpha-anomer of Gal at nonreducing end and most complementary to Galalpha1-->6Glc. As for the combining site of PA-IL toward the beta-anomer, the size is assumed to be less than that of Gal; carbon-6 in the pyranose form is essential, and hydrophobic interaction is important for binding.
Sigma-Aldrich
Mucin from porcine stomach, Type III, bound sialic acid 0.5-1.5 %, partially purified powder
Sigma-Aldrich
Mucin from porcine stomach, Type II