Merck
  • Identification and quantification of alkene-based drilling fluids in crude oils by comprehensive two-dimensional gas chromatography with flame ionization detection.
Identification and quantification of alkene-based drilling fluids in crude oils by comprehensive two-dimensional gas chromatography with flame ionization detection.
Journal of chromatography. A (2007-03-23)
Christopher M Reddy, Robert K Nelson, Sean P Sylva, Li Xu, Emily A Peacock, Bhavani Raghuraman, Oliver C Mullins
ABSTRACT
Comprehensive two-dimensional gas chromatography with flame ionization detection (GC x GC-FID) was used to measure alkene-based drilling fluids in crude oils. Compared to one-dimensional gas chromatography, GC x GC-FID is more robust for detecting alkenes due to the increased resolution afforded by second dimension separations. Using GC x GC-FID to analyze four oil samples from one reservoir contaminated with the same drilling fluid, C(15), C(16), C(17), C(18) and C(20) alkenes were identified. The drilling fluid that contaminated these samples also differed from another commercially obtained fluid, which only contained C(16) and C(18) alkenes. These results should motivate the petroleum industry to consider GC x GC-FID for measuring drilling fluids.
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Pricing and availability is not currently available.