Merck
  • Low-field MR arthrography of the shoulder joint: technique, indications, and clinical results.
Low-field MR arthrography of the shoulder joint: technique, indications, and clinical results.
European radiology (2003-02-25)
K-F Kreitner, R Loew, M Runkel, J Zöllner, M Thelen
ABSTRACT
In the age of cost containment and urgent reductions in health care expenditures, new options have to be explored to satisfy both diagnostic requirements and economic limitations. The introduction of low-field MR systems for assessment of joint disorders seemed to be an option for lower costs. The purpose of this article is to summarize available experiences with low-field MR arthrography of the glenohumeral joint with respect to image quality and diagnostic accuracy in detecting labral and rotator cuff lesions. Up to now, there has been only a limited number of studies available dealing with low-field MR arthrography of the glenohumeral joint. They reveal that, despite a minor image quality in comparison with high-field imaging, low-field MR arthrography of the shoulder allows for sufficient evaluation of intra- and extra-articular structures in the detection of major abnormalities such as glenohumeral instability or rotator cuff disease. Furthermore, open-configured MR scanners enable kinematic studies: Besides the analysis of normal motion, pathological findings in patients with instabilities and impingement syndrome can be delineated. They further offer the possibility for performing MR imaging-guided arthrography of the shoulder. This was first described using an open C-arm scanner with a vertically oriented magnetic field so that MR arthrography may be performed in one setting.
Product Description
Iotrolan for system suitability, European Pharmacopoeia (EP) Reference Standard
Iotrolan, European Pharmacopoeia (EP) Reference Standard