Merck
  • Characterization of cross-linked hydroxycinnamic acid amides isolated from potato common scab lesions.
Characterization of cross-linked hydroxycinnamic acid amides isolated from potato common scab lesions.
Phytochemistry (2005-09-03)
Russell R King, Larry A Calhoun
ABSTRACT
Four feruloyl amides, N-trans-feruloyloctopamine (1), N-cis-feruloyloctopamine (2), N-trans-feruloyltyramine (3), N-cis-feruloyltyramine (4), a cross-linked N-trans-feruloyltyramine dimer (5), and a cross-linked N-cis-feruloyltyramine dimer (6) were isolated from potato common scab lesions. The compounds were purified by TLC and characterized by a combination of (1)H and (13)C NMR spectroscopic techniques. The presence of an accompanying minor complex of cross-linked dimers containing both feruloyltyramines and feruloyloctopamines was also demonstrated. This is the first characterization of cross-linked hydroxycinnamic acid amides associated with wound healing in potato (Solanum tuberosum) tubers.
Sigma-Aldrich
Moupinamide