Merck
  • Preparative isolation of hydrolysable tannins chebulagic acid and chebulinic acid from Terminalia chebula by high-speed counter-current chromatography.
Preparative isolation of hydrolysable tannins chebulagic acid and chebulinic acid from Terminalia chebula by high-speed counter-current chromatography.
Journal of separation science (2006-08-23)
Quanbin Han, Jingzheng Song, Chunfeng Qiao, Lina Wong, Hongxi Xu
ABSTRACT
As a chromatographic column, the high-speed counter-current chromatography system was equipped with a preparative HPLC series, enabling the successful isolation of hydrolysable tannins from the fruits of Terminalia chebula, a traditional Chinese medicine. The two-phase solvent system was composed of n-hexane-ethyl acetate-methanol-water (1:20:1:20 v/v). As a result, 33.2 mg chebulagic and 15.8 mg chebulinic acids were obtained in one step from 300 mg of crude extract. Their purities were determined by HPLC to be 95.3 and 96.1%, respectively. The chemical structures were identified by their MS and 1H NMR spectra.
Product Description
Supelco
Chebulagic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Chebulinic acid, ≥85% (LC/MS-ELSD)
Pricing and availability is not currently available.