Merck
  • Dose-dependent hemodynamic effects of propofol induction following brotizolam premedication in hypertensive patients taking angiotensin-converting enzyme inhibitors.
Dose-dependent hemodynamic effects of propofol induction following brotizolam premedication in hypertensive patients taking angiotensin-converting enzyme inhibitors.
Journal of clinical anesthesia (2010-04-20)
Marian Weisenberg, Daniel I Sessler, Monica Tavdi, Muzikant Gleb, Tiberiu Ezri, Jarrod E Dalton, Michael Protianov, Reuven Zimlichmann
ABSTRACT
To determine a propofol dose that minimizes hemodynamic changes on induction of anesthesia in patients chronically taking angiotensin-converting enzyme inhibitors (ACEIs). Prospective, randomized trial. Operating room of a university-affiliated general hospital. 88 ASA physical status II and II hypertensive patients chronically taking ACEIs, scheduled for elective abdominal surgery with general anesthesia. Patients were premedicated with brotizolam and anesthesia was induced with propofol, fentanyl, and rocuronium; anesthesia was then maintained with isoflurane. Patients were randomly assigned to undergo anesthetic induction with propofol in doses of 1.3, 1.6, 2.0, or 2.3 mg/kg. Oscillometric blood pressure and heart rate were evaluated at one-minute intervals during the first 10 minutes of anesthesia. End-tidal isoflurane concentrations were also recorded. Episodes of hypertension, tachycardia, bradycardia, or hypotension (defined as > 30% of baseline values) were managed per protocol with esmolol, atropine, phenylephrine, or ephedrine. Administration of any of these drugs was considered a pharmacological intervention. After adjusting for covariables in a model assuming a linear relationship between dose and log-response, each propofol dose increase of 0.3 mg/kg was associated with a 31% increase in mean number of hypotensive/bradycardic episodes requiring interventions (95% confidence intervals of +5% and +65%; P = 0.018). Based on our model, a dose of 1.3 mg/kg resulted in the fewest number of pharmacological interventions. In patients chronically taking ACEIs, low doses of propofol reduce hemodynamic instability.