Merck
  • Treatment with n-3 polyunsaturated fatty acids reverses endothelial dysfunction and oxidative stress in experimental menopause.
Treatment with n-3 polyunsaturated fatty acids reverses endothelial dysfunction and oxidative stress in experimental menopause.
The Journal of nutritional biochemistry (2012-11-20)
Gianluca Gortan Cappellari, Pasquale Losurdo, Sara Mazzucco, Emiliano Panizon, Mitja Jevnicar, Loredana Macaluso, Bruno Fabris, Rocco Barazzoni, Gianni Biolo, Renzo Carretta, Michela Zanetti
ABSTRACT
Menopause is associated with endothelial dysfunction and oxidative stress. In this condition, reduced n-3 polyunsaturated fatty acids (n-3 PUFAs) contribute to cardiovascular disease. We investigated whether treatment with n-3 PUFA reverses endothelial dysfunction and oxidative stress in experimental menopause. Thirty female rats underwent either sham-surgery or bilateral ovariectomy or bilateral ovariectomy+oral n-3 PUFA (0.8 g kg(-1) day(-1) for 2 months). Ovariectomy caused endothelial dysfunction to acetylcholine, which was reversed by superoxide scavenger Tiron. Erythrocyte membrane lipid composition was characterized by reduced n-3 PUFA total content and omega-3 index, and by concomitant increase in n-6:n-3 PUFA ratio. Ovariectomy-related oxidative stress, demonstrated by both enhanced superoxide production and 3-nitrotyrosine expression in aorta, was associated with increased nicotinamide adenine dinucleotide phosphate (NADPH) oxidase subunit NOX-4 protein expression. Endothelial nitric oxide synthase (eNOS) functional inhibition by l-NG-nitroarginine methyl ester, protein expression and activity did not change. In ovariectomized rats, treatment with n-3 PUFA increased n-3 PUFA total content and omega-3 index and decreased n-6:n-3 PUFA ratio in erythrocyte membrane, reversed vascular oxidative stress, endothelial dysfunction, aortic 3-nitrotyrosine and markedly lowered NOX-4 protein expression; eNOS protein expression also increased, paralleled by reversal of inhibitory binding to Caveolin-1, while ex-vivo functional inhibition and NOS synthesis were unchanged. These findings demonstrate in vivo a therapeutic benefit of n-3 PUFA on menopause-associated endothelial dysfunction by reversal of alterations in membrane lipid composition induced by ovariectomy and by reduction of vascular oxidative stress. In this setting they also identify NOX-4 as a potential target to reduce oxidative stress-mediated vascular complications.