Merck
  • Sublethal effects of the antibiotic tylosin on estuarine benthic microalgal communities.
Sublethal effects of the antibiotic tylosin on estuarine benthic microalgal communities.
Marine pollution bulletin (2013-02-13)
James L Pinckney, Isaac M Hagenbuch, Richard A Long, Charles R Lovell
ABSTRACT
Pharmaceuticals are common chemical contaminants in estuaries receiving effluent from wastewater and sewage treatment facilities. The purpose of this research was to examine benthic microalgal (BMA) community responses to sublethal exposures to tylosin, a common and environmentally persistent antibiotic. Bioassays, using concentrations of 0.011-218 μmol tylosin l(-1), were performed on intertidal muddy sediments from North Inlet Estuary, SC. Exposure to tylosin resulted in a reduction in total BMA biomass and primary productivity. Furthermore, exposure seemed to retard diatom growth while having a minimal effect on cyanobacteria biomass. Estuarine systems receiving chronic inputs of trace concentrations of tylosin, as well as other antibiotics, may experience significant reductions in BMA biomass and primary productivity. Given the well-documented role of BMA in the trophodynamics of estuaries, these impacts will likely be manifested in higher trophic levels with possible impairments of the structure and function of these sensitive systems.