Merck
Anaphylaxislike cholinergic urticaria.
Canadian family physician Medecin de famille canadien (2013-07-16)
Darío Antolín-Amérigo, Petruta Cristina Vlaicu, Belén De La Hoz Caballer, Moisés Sánchez Cano
Product Description
Montelukast sodium, European Pharmacopoeia (EP) Reference Standard