Merck
  • The phytotherapeutic agent, eviprostat, suppresses stromal proliferation and inflammation even after establishment of nonbacterial prostatitis in the rat prostate.
The phytotherapeutic agent, eviprostat, suppresses stromal proliferation and inflammation even after establishment of nonbacterial prostatitis in the rat prostate.
Urology (2014-03-04)
Shinsuke Shibuya, Zhang Xia, Mikio Sugimoto, Nobufumi Ueda, Reiji Haba, Yoshiyuki Kakehi
ABSTRACT
To evaluate the effect of phytotherapeutic agent, Eviprostat, administered after the establishment of nonbacterial prostatitis (NBP) on the stroma-to-epithelium ratio (S/E ratio), inflammatory scores, tissue macrophage infiltration, and cytokines and chemokines levels in prostate tissue and urine. Ten-month-old male Wistar rats were castrated and exposed to 17-beta-isomer of estradiol for 30 days to induce NBP. Twenty-five NBP rats were divided into 5 groups: (1) NBP (0) rats sacrificed immediately after the establishment of NBP; (2,3) NBP (30)/control (CTL) and NBP (30)/Eviprostat (EVI) rats fed without or with 0.1% Eviprostat under estradiol-free for 30 days, respectively; and (4,5) NBP (60)/CTL and NBP (60)/EVI rats fed without or with 0.1% Eviprostat under estradiol-free for 60 days, respectively. The S/E ratio, inflammatory scores, and the number of macrophage infiltration in the prostate were assessed. Concentrations of cytokines and chemokines in prostatic tissue and urine were measured by enzyme-linked immunosorbent assay. The S/E ratio was significantly increased with time until 60 days under estradiol-free condition (P <.001). The S/E ratio and the inflammatory scores in NBP (60)/EVI was significantly lower than that of NBP (60)/CTL (P <.001, and P = .022, respectively). The mean tissue concentration of chemokine ligand 2 (CCL2)/monocyte chemoattractant protein-1 (MCP-1) in NBP (60)/CTL was significantly higher than that in NBP (0) (P = .016), whereas, there was no difference between NBP (60)/EVI and NBP (0). Furthermore, urinary CCL2/MCP-1 was significantly decreased in NBP (60)/EVI as compared with NBP (0) (P = .028). Eviprostat suppresses the stromal proliferation and inflammation in the rat prostate after the establishment of NBP at least partly owing to inhibitory effect on CCL2/MCP-1 production in the prostate.
Product Description
Etamsylate, European Pharmacopoeia (EP) Reference Standard