Merck
  • Micellar electrokinetic chromatography as a fast screening method for the determination of the doping agents furosemide and piretanide in urine.
Micellar electrokinetic chromatography as a fast screening method for the determination of the doping agents furosemide and piretanide in urine.
Journal of chromatography. B, Biomedical sciences and applications (1997-01-10)
S P Lalljie, M B Barroso, D Steenackers, R M Alonso, R M Jiménez, P Sandra
ABSTRACT
The possibility of using micellar electrokinetic chromatography for the screening of the loop diuretics piretanide and furosemide in urine was studied. A fast and simple method with good repeatability is described. The method was applied to urine samples collected from a healthy volunteer after oral administration of therapeutic doses of each compound. Positive identification in the urine matrix was possible through recording diode array spectra.
Product Description
Piretanide for system suitability, European Pharmacopoeia (EP) Reference Standard
Piretanide, European Pharmacopoeia (EP) Reference Standard