Merck
3D reconstruction of high-resolution STED microscope images.
Microscopy research and technique (2008-06-03)
Annedore Punge, Silvio O Rizzoli, Reinhard Jahn, Jan Dominik Wildanger, Lars Meyer, Andreas Schönle, Lars Kastrup, Stefan W Hell
ABSTRACT
Tackling biological problems often involves the imaging and localization of cellular structures on the nanometer scale. Although optical super-resolution below 100 nm can be readily attained with stimulated emission depletion (STED) and photoswitching microscopy methods, attaining an axial resolution <100 nm with focused light generally required the use of two lenses in a 4Pi configuration or exceptionally bright photochromic fluorophores. Here, we describe a simple technical solution for 3D nanoscopy of fixed samples: biological specimens are fluorescently labeled, embedded in a polymer resin, cut into thin sections, and then imaged via STED microscopy with nanoscale resolution. This approach allows a 3D image reconstruction with a resolution <80 nm in all directions using available state-of-the art STED microscopes.
Sigma-Aldrich
Atto 633, BioReagent, suitable for fluorescence
Sigma-Aldrich
Phalloidin–Atto 532, suitable for fluorescence, ≥90.0% (HPLC)
Supelco
Atto 647N maleimide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647N azide, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 532 maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)
Sigma-Aldrich
Atto 425, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 647N amine, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 647N alkyne, BioReagent, suitable for fluorescence
Sigma-Aldrich
Phalloidin–Atto 647N, BioReagent, suitable for fluorescence, ≥80% (HPLC)
Sigma-Aldrich
Atto 425-Biotin, BioReagent, suitable for fluorescence, ≥95.0% (HPCE)
Sigma-Aldrich
Atto 425-Streptavidin, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 647N DOPE, suitable for fluorescence
Sigma-Aldrich
Phalloidin–Atto 390, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 390 azide, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
Atto 633 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647N-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Atto 647N Protein Labeling Kit, BioReagent, suitable for fluorescence
Sigma-Aldrich
Phalloidin–Atto 590, suitable for fluorescence, ≥90%
Sigma-Aldrich
Atto 425 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 590-Biotin, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Sigma-Aldrich
Atto 590-Streptavidin, BioReagent, suitable for fluorescence
Supelco
Atto 390 NHS ester, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 647N NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 532, BioReagent, suitable for fluorescence, ≥90% (HPCE)
SAFC
Atto 465 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 647N-Streptavidin, BioReagent, suitable for fluorescence
Sigma-Aldrich
Phalloidin–Atto 425, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 532 NHS ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Atto 532 DOPE, suitable for fluorescence