Merck
  • High-yield production of a low-temperature-active polygalacturonase for papaya juice clarification.
High-yield production of a low-temperature-active polygalacturonase for papaya juice clarification.
Food chemistry (2013-07-23)
Tao Tu, Kun Meng, Yingguo Bai, Pengjun Shi, Huiying Luo, Yaru Wang, Peilong Yang, Yuhong Zhang, Wei Zhang, Bin Yao
ABSTRACT
A novel endo-polygalacturonase (endo-PG I) from Achaetomium sp. Xz8 was identified, overexpressed in Pichia pastoris, and characterized in this report. Recombinant endo-PG I is distinguished from other enzyme counterparts by its high activity towards polygalacturonic acid (49,934 U/ml) and high yield in the 15-l fermentor (2.13 g/l). It exhibits optimal activity at 45 °C and remained active over a broad temperature range of 0-80 °C. Distinct from most fungal polygalacturonases that have acidic pH optima, endo-PG I is optimally active at pH 6, similar to the pH of fresh papaya juice (5.7). Endo-PG I alone reduced the viscosity of papaya juice by 17.6%, and increased its transmittance by 59.1%. When combined with a commercial pectin methylesterase, it showed much higher efficiency with a synergy degree of more than 1.25. All these favourable enzymatic properties make endo-PG I attractive for potential applications in the juice industry.
Sigma-Aldrich
Pectinase from Aspergillus niger, BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, suitable for plant cell culture, lyophilized powder
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, lyophilized powder, ≥0.3 units/mg solid
Sigma-Aldrich
Pectinase from Rhizopus sp., powder, 400-800 units/g solid