Merck
  • No impact of high-dose cytarabine and asparaginase as early intensification with intermediate-risk paediatric acute lymphoblastic leukaemia: results of randomized trial TCCSG study L99-15.
No impact of high-dose cytarabine and asparaginase as early intensification with intermediate-risk paediatric acute lymphoblastic leukaemia: results of randomized trial TCCSG study L99-15.
British journal of haematology (2013-10-30)
Motohiro Kato, Katsuyoshi Koh, Atsushi Manabe, Tomohiro Saito, Daisuke Hasegawa, Keiichi Isoyama, Akitoshi Kinoshita, Miho Maeda, Yuri Okimoto, Michiko Kajiwara, Takashi Kaneko, Kanji Sugita, Akira Kikuchi, Masahiro Tsuchida, Akira Ohara
ABSTRACT
The Tokyo Children's Cancer Study Group conducted a randomized controlled study to evaluate the effect of experimental early intensification using high-dose cytarabine and L-asparaginase in paediatric intermediate-risk (IR) acute lymphoblastic leukaemia (ALL). A total of 310 IR ALL patients were randomized to receive either experimental early intensification (n = 156) or standard early intensification including standard-dose cytarabine arm (n = 154) after induction therapy. The experimental arm consisted of high-dose cytarabine and L-asparaginase, while the standard arm consisted of standard-dose cytarabine, oral 6-mercaptopurine and cyclophosphamide. The probabilities of event-free survival at 8 years in the experimental and standard arms were 72·3 ± 3·7% and 77·5 ± 3·5%, respectively (P = 0·32). The 8-year overall survival rates for these two arms were 85·0 ± 3·0% and 86·9 ± 2·8%, respectively (P = 0·72). The frequency of infectious events was significantly higher in the experimental arm (66·4%) than in the standard arm (24·6%) (P < 0·001). In conclusion, experimental early intensification including high-dose cytarabine followed by L-asparaginase had no advantage over standard early intensification in paediatric IR ALL patients.
Sigma-Aldrich
Asparaginase from Escherichia coli, lyophilized powder, 100-300 units/mg protein (biuret)