Merck
  • Tear fluid extracellular DNA: diagnostic and therapeutic implications in dry eye disease.
Tear fluid extracellular DNA: diagnostic and therapeutic implications in dry eye disease.
Investigative ophthalmology & visual science (2013-11-21)
Sapna Tibrewal, Joy Sarkar, Sarmad H Jassim, Sonal Gandhi, Snehal Sonawane, Shweta Chaudhary, Yong-Soo Byun, Yair Ivanir, Joelle Hallak, John H Horner, Martin Newcomb, Sandeep Jain
ABSTRACT
To determine the abundance of extracellular DNA (eDNA) in tear fluid of patients with dry eye disease (DED) and to report clinical outcomes after DNase I eyedrops use to reduce excessive tear fluid eDNA. Tear fluid was collected from healthy control subjects and patients with DED. The eDNA abundance was determined with the PicoGreen dye assay. The DED symptoms and clinical signs were recorded and correlated with eDNA abundance. Two patients with DED having excessive eDNA in tear fluid were treated with DNase I eyedrops. The PicoGreen dye assay measures tear fluid eDNA abundance after a 2-minute incubation time. With longer incubations, admixed cells also contribute to eDNA measurements. The mean (SE) eDNA abundance in healthy control subjects' tear fluid was 1.4 (0.2) μg/mL. The mean (SE) eDNA abundance in tear fluid of patients with nonautoimmune DED, autoimmune DED, and graft versus host disease was significantly higher: the values were 2.9 (0.6), 5.2 (1.2), and 9.1 (2.3) μg/mL, respectively (P < 0.05). In most of these patients, the PicoGreen dye kinetic assay of tear fluid showed an increase in fluorescence signal due to the presence of viable cells in tear fluid. Tear fluid eDNA had the best correlation with corneal Rose Bengal staining (r = 0.55). Treatment of patients having DED with DNase I eyedrops reduced eDNA abundance, abrogated signal increase, and improved comfort. Excessive eDNA is present in tear fluid of patients with dry eyes. A novel therapeutic approach for managing DED may be to measure eDNA abundance in tear fluid with the PicoGreen dye assay and reduce excessive amounts with DNase I eyedrops.
Sigma-Aldrich
Deoxyribonuclease I RNase-free solution from bovine pancreas
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
DNase I, Amplification Grade
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, lyophilized powder, RNAse and protease, free