Merck
  • Trypsin and rosmarinic acid reduce the toxicity of Micrurus fulvius venom in mice.
Trypsin and rosmarinic acid reduce the toxicity of Micrurus fulvius venom in mice.
Clinical toxicology (Philadelphia, Pa.) (2014-01-15)
J L Parker-Cote, D P O'Rourke, S N Miller, K L Brewer, M D Rosenbaum, W J Meggs
ABSTRACT
CONTEXT. Antivenom is expensive and not always available, so alternative treatments are being investigated. OBJECTIVE. The efficacy of trypsin or rosmarinic acid (RA) in treating Micrurus fulvius in a murine model is determined. MATERIALS AND METHODS. randomized controlled blinded study. Fifty mice (20-30 g). Study groups: Intraperitoneal injections of: 1) 2 mg/kg M. fulvius venom (approximately twice the LD50 for mice; n = 10); 2) 2 mg/kg M. fulvius venom incubated in vitro for 1 h prior to injection with RA at a 1:10 ratio (n = 17); 3) 2 mg/kg M. fulvius venom incubated in vitro for 1 h prior to injection with 1 mg of trypsin (n = 17); 4)1 mg trypsin IP without venom (n = 3); and 5) RA IP without venom (n = 3). time to toxicity (respiratory distress (< 25 breaths/min.), loss of spontaneous locomotor activity, or inability to upright self). Time to toxicity using Tukey-Kramer HSD; Survival to 4, 6, and 12 h using Chi-square analysis. RESULTS. Onset of toxicity: venom + saline, 120.3 + 64.4 min; venom + rosmarinic acid, 238.1 ± 139.2 min (p = 0.15 relative to venom + saline); venom + trypsin, 319.7 + 201.0 min (p = 0.007 relative to venom + saline). Venom + trypsin but not venom + RA survival to 4 h was significant compared to venom + saline (p = 0.023). Two mice in the venom + trypsin group and one mouse in the venom + RA group survived to 12 h. Mice receiving trypsin without venom or RA without venom survived to 12 h without toxicity. Discussion. This work suggests that trypsin and RA may have efficacy in treatment M. fulvius envenomation. CONCLUSION. In vitro neutralization of M. Fulvius venom by trypsin justifies progressing to an in vivo model in future studies.
Sigma-Aldrich
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
Sigma-Aldrich
Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Sigma-Aldrich
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
Trypsin from porcine pancreas, ~1500 U/mg
Sigma-Aldrich
Trypsin from porcine pancreas, tablet, 1 mg tablet
Sigma-Aldrich
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid